missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. aeruginosa Regulatory protein RhlR Protein
A DNA sequence encoding the Pseudomonas aeruginosa Regulatory protein RhlR, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | NP_252167.1 |
---|---|
Gene ID (Entrez) | 878968 |
Name | P. aeruginosa Regulatory protein RhlR Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16017800
|
enQuireBio™
QP7228-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037800
|
enQuireBio™
QP7228-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027800
|
enQuireBio™
QP7228-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047800
|
enQuireBio™
QP7228-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057800
|
enQuireBio™
QP7228-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067800
|
enQuireBio™
QP7228-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_252167.1 | |
P. aeruginosa Regulatory protein RhlR Protein | |
Not determined. | |
Full Length | |
P. aeruginosa | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
878968 | |
Research Use Only | |
rhlR | |
Recombinant Protein | |
E. coli | |
MRNDGGFLLW WDGLRSEMQP IHDSQGVFAV LEKEVRRLGF DYYAYGVRHT IPFTRPKTEV HGTYPKAWLE RYQMQNYGAV DPAILNGLRS SEMVVWSDSL FDQSRMLWNE ARDWGLCVGA TLPIRAPNNL LSVLSVARDQ QNISSFEREE IRLRLRCMIE LLTQKLTDLE HPMLMSNPVC LSHREREILQ WTADGKSSGE IAIILSISES TVNFHHKNIQ KKFDAPNKTL AAAYAAALGL I | |
Greater than 84.5% as determined by SDS-PAGE. |