missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pig Epididymal secretory glutathione peroxidase Protein

A DNA sequence encoding the Sus scrofa (Pig) Epididymal secretory glutathione peroxidase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6117-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16080930

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_999051.1
Epididymal secretory glutathione peroxidase Protein
Research Use Only
GPX5
Recombinant Protein
Yeast
NSNLEKMDCY KDVTGTIYDY DAFTLNGNEH IQFKQYAGKH VLFVNVATYC GLTAQYPELN TLQEELKPFG LVVLGFPCNQ FGKQEPGENS EILLGLKYVR PGGGYVPNFQ LFEKGDVNGE KEQKVFTFLK HSCPHPSELI GSIGYISWEP IRVHDIRWNF EKFLVGPDGV PVMRWVHETP ISTVKSDILA YLKQFKTE
Greater than 90% as determined by SDS-PAGE.
396920
50 μg
Not determined.
Full Length of Mature Protein
Pig
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers