missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Erythropoietin / EPO Protein
A DNA sequence encoding the Sus scrofa (Pig) Erythropoietin / EPO, was expressed in the hosts and tags indicated.
287.00 € - 3175.00 €
Specifications
Accession Number | NP_999299.1 |
---|---|
Gene ID (Entrez) | 397249 |
Name | Erythropoietin / EPO Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15919189
|
enQuireBio™
QP5990-YE-100UG |
100 μg |
880.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959189
|
enQuireBio™
QP5990-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929189
|
enQuireBio™
QP5990-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15949189
|
enQuireBio™
QP5990-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15969189
|
enQuireBio™
QP5990-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15939189
|
enQuireBio™
QP5990-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_999299.1 | |
Erythropoietin / EPO Protein | |
Not determined. | |
Full Length | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |
397249 | |
Research Use Only | |
EPO | |
Recombinant Protein | |
Yeast | |
APPRLICDSR VLERYILEAK EGENATMGCA ESCSFSENIT VPDTKVNFYA WKRMEVQQQA MEVWQGLALL SEAILQGQAL LANSSQPSEA LQLHVDKAVS GLRSLTSLLR ALGAQKEAIP LPDASPSSAT PLRTFAVDTL CKLFRNYSNF LRGKLTLYTG EACRRRDR | |
Greater than 90% as determined by SDS-PAGE. |