missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pig Erythropoietin / EPO Protein

A DNA sequence encoding the Sus scrofa (Pig) Erythropoietin / EPO, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5990-ma-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989179

Quantity Price
1 1407.27 € / 100µg
3 1323.54 € / 100µg
Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

NP_999299.1
Erythropoietin / EPO Protein
100 μg
Not determined.
Full Length
Pig
His
Tris-based buffer, 50% glycerol
397249
Greater than 90% as determined by SDS-PAGE.
Research Use Only
EPO
Recombinant Protein
Mammal
APPRLICDSR VLERYILEAK EGENATMGCA ESCSFSENIT VPDTKVNFYA WKRMEVQQQA MEVWQGLALL SEAILQGQAL LANSSQPSEA LQLHVDKAVS GLRSLTSLLR ALGAQKEAIP LPDASPSSAT PLRTFAVDTL CKLFRNYSNF LRGKLTLYTG EACRRRDR
Videos
SDS
Documents
Certificates