missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig IL-8 / CXCL8 Protein
Recombinant Protein
107.00 € - 4060.00 €
Specifications
Accession Number | NP_999032.1 |
---|---|
Gene ID (Entrez) | 396880 |
Name | IL-8 / CXCL8 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15916339
|
enQuireBio™
QP5522-5UG |
5 μg |
107.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996329
|
enQuireBio™
QP5522-20UG |
20 μg |
252.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976329
|
enQuireBio™
QP5522-100UG |
100 μg |
650.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15906339
|
enQuireBio™
QP5522-500UG |
500 μg |
2490.00 €
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986329
|
enQuireBio™
QP5522-1MG |
1 mg |
4060.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_999032.1 | |
IL-8 / CXCL8 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
ARVSAELRCQ CINTHSTPFH PKFIKELRVI ESGPHCENSE IIVKLVNGKE VCLDPKEKWV QKVVQIFLKR TEKQQQQQ | |
Greater than 95% as determined by SDS-PAGE. |
396880 | |
Research Use Only | |
CXCL8 | |
Pig | |
Untagged | |
5 mM sodium phosphate, 20 mM sodium chloride, pH 7.5 |