missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Trypsin Protein
A DNA sequence encoding the Sus scrofa (Pig) Trypsin, was expressed in the hosts and tags indicated.
352.00 € - 3900.00 €
Specifications
Name | Trypsin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Protein Length | Full Length of Mature Protein |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048151
|
enQuireBio™
QP9350-YE-100UG |
100 μg |
1050.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16068151
|
enQuireBio™
QP9350-YE-1MG |
1 mg |
3900.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058151
|
enQuireBio™
QP9350-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078151
|
enQuireBio™
QP9350-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088151
|
enQuireBio™
QP9350-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098151
|
enQuireBio™
QP9350-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Trypsin Protein | |
Not determined. | |
Recombinant Protein | |
Yeast | |
IVGGYTCAAN SIPYQVSLNS GSHFCGGSLI NSQWVVSAAH CYKSRIQVRL GEHNIDVLEG NEQFINAAKI ITHPNFNGNT LDNDIMLIKL SSPATLNSRV ATVSLPRSCA AAGTECLISG WGNTKSSGSS YPSLLQCLKA PVLSDSSCKS SYPGQITGNM ICVGFLEGGK DSCQGDSGGP VVCNGQLQGI VSWGYGCAQK NKPGVYTKVC NYVNWIQQTI AAN | |
Greater than 90% as determined by SDS-PAGE. |
Research Use Only | |
Full Length of Mature Protein | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |