missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine Enterokinase Protein
A cDNA sequence encoding the Enterokinase was constructed and used to recombinantly synthesize the protein.
373.00 € - 2175.00 €
Specifications
Gene ID (Entrez) | 397152 |
---|---|
Name | Enterokinase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | TMPRSS15 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15968378
|
enQuireBio™
QP11791-1000IU |
1000 IU |
2175.00 €
1000 units |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978378
|
enQuireBio™
QP11791-100IU |
100 IU |
373.00 €
100 units |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988378
|
enQuireBio™
QP11791-500IU |
500 IU |
1305.00 €
500 units |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
397152 | |
Research Use Only | |
TMPRSS15 | |
Porcine | |
Untagged | |
2 IU/microL, 50mM Tris-HCl, pH 8.0, 0.5M NaCl and 50% glycerol. |
Enterokinase Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Porcine | |
MGSKRIIPSR HRSLSTYEVM FTALFAILMV LCAGLIAVSW LTIKGSEKDA ALGKSHEARG TMKITSGVTY NPNLQDKLSV DFKVLAFDIQ QMIGEIFQSS NLKNEYKNSR VLQFENGSVI VIFDLLFAQW VSDENIKEEL IQGIEANKSS QLVAFHIDVN SIDITESLEN YSTTSPSTTS DKLTTSSPPA TPGNVSIECL PGSRPCADAL KCIAVDLFCD GELNCPDGSD EDSKICATAC DGKFLLTESS GSFDAAQYPK LSEASVVCQW IIRVNQGLSI ELNFSYFNTY SMDVLNIYEG VGSSKILRAS LWLMNPGTIR IFSNQVTVTF LIESDENDYI GFNATYTAFN STELNNDEKI NCNFEDGFCF WIQDLNDDNE WERIQGTTFP PFTGPNFDHT FGNASGFYIS TPTGPGGRQE RVGLLSLPLE PTLEPVCLSF WYYMYGENVY KLSINISNDQ NIEKIIFQKE GNYGENWNYG QVTLNETVEF KVAFNAFKNQ FLSDIALDDI SLTYGICNVS LYPEPTLVPT SPPELPTDCG GPFELWEPNT TFTSMNFPNN YPNQAFCVWN LNAQKGKNIQ LHFEEFDLEN IADVVEIRDG EEDDSLLLAV YTGPGPVEDV FSTTNRMTVL FITNDALTKG GFKANFTTGY HLGIPEPCKE DNFQCENGEC VLLVNLCDGF SHCKDGSDEA HCVRFLNGTA NNSGLVQFRI QSIWHTACAE NWTTQTSDDV CQLLGLGTGN SSMPFFSSGG GPFVKLNTAP NGSLILTASE QCFEDSLILL QCNHKSCGKK QVAQEVSPKI VGGNDSREGA WPWVVALYYN GQLLCGASLV SRDWLVSAAH CVYGRNLEPS KWKAILGLHM TSNLTSPQIV TRLIDEIVIN PHYNRRRKDS DIAMMHLEFK VNYTDYIQPI |