missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine Leptin Protein
A cDNA sequence encoding the Leptin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10752-100ug
Additional Details : Weight : 0.01000kg
Specifications
Leptin Protein | |
Research Use Only | |
Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. | |
Porcine | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.02% NaHCO3. |
100 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MRCGPLCRFL WLWPYLSYVE AVPIWRVQDD TKTLIKTIVT RISDISHMQS VSSKQRVTGL DFIPGLHPVL SLSKMDQTLA IYQQILTSLP SRNVIQISND LENLRDLLHL LASSKSCPLP QARALETLES LGGVLEASLY STEVVALSRL QGALQDMLRQ LDLSPGC | |
Greater than 98.0% as determined by:(a) Gel filtration analysis. (b) Analysis by SDS-PAGE. |