missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine Trypsin Protein
A cDNA sequence encoding the Trypsin was constructed and used to recombinantly synthesize the protein.
131.00 € - 1305.00 €
Specifications
Name | Trypsin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Product Type | Recombinant Protein |
Cross Reactivity | Porcine |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985498
|
enQuireBio™
QP10897-1MG |
1 mg |
131.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995498
|
enQuireBio™
QP10897-2.5MG |
2.5 mg |
203.00 €
2.50mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15905508
|
enQuireBio™
QP10897-50MG |
50 mg |
1305.00 €
50mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Trypsin Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Porcine | |
Untagged | |
The Porcine Trypsin (2.98 mg/ml) is formulated with 1mM HCl and 20mM CaCl2, pH 3. |
Research Use Only | |
Recombinant Protein | |
P. pastoris | |
VGGYTCAANSIPYQVSLNSGSHFCGGSLINSQWVVSAAHCYKSRIQVRLGEHNI DVLEGNEQFINAAKIITHPNFNGNTLDNDIMLIKLSSPATLNSRVATVSLPRSCA AAGTECLISGWGNTKSSGSSYPSLLQCLKAPVLSDSSCKSSYPGQITGNMICVGF LEGGKDSCQGDSGGPVVCNGQLQGIVSWGYGCAQKNKPGVYTKVCNYVNWI QQTIAAN |