missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit E-Selectin / CD62e / SELE Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) E-Selectin / CD62e / SELE, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9384-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001075781.1 | |
E-Selectin / CD62e / SELE Protein | |
Research Use Only | |
SELE | |
Recombinant Protein | |
Yeast | |
WTYHFSAENM TYDEASAYCQ QNYTHLVAIQ NKEEIDYLNS ILDYSPSYYW IGIRKVNNVW IWVGTHKPLT EGAKNWAPGE PNNKQNNEDC VEIYIKRPKD TGMWNDERCS KKKLALCYTA ACTEASCSGH GECIETINNY SCKCYPGFSG LKCEQVVTCE AQVQPQHGSL NCTHPLGNFS YNSSCSVSCE RGYLPSSTET TWCTSSGEWS APPATCKVVE CDTMGKPANG DVKCSPSQGS APWNTTCTFD CEEGFTLLGA RSLQCTSSGS WDNEKPTCKA VSCDTIHHPQ NGSVSCSNSS EGKFTFRSSC NFTCEENFLL RGPAQVECTA QGQWTQQAPV CEAVKCDPVH TLEDGFVKCT HPHTGEFTYK SSCTFNCREG FELHGSAQLE CTSQGQWAQE LPSCQVVQCP SLAVLGKTNV SCSGEPVFGT VCNFACPEGW TLNGSAALMC GAEGQWSGML PTCEEPIASN VP | |
Greater than 90% as determined by SDS-PAGE. |
100009151 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Rabbit | |
His | |
Tris-based buffer, 50% glycerol |