missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rabbit GH Protein

A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11963-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988938

Quantity Price
1 114.77 € / 10µg
10 107.33 € / 10µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

GH Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Rabbit
Untagged
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3.
Greater than 97.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MAAGSWTAGL LAFALLCLPW PQEASAFPAM PLSSLFANAV LRAQHLHQLA ADTYKEFERA YIPEGQRYSI QNAQAAFCFS ETIPAPTGKD EAQQRSDMEL LRFSLLLIQS WLGPVQFLSR AFTNTLVFGT SDRVYEKLKD LEEGIQALMR ELEDGSPRVG QLLKQTYDKF DTNLRGDDAL LKNYGLLSCF KKDLHKAETY LRVMKCRRFV ESSCVF
Videos
SDS
Documents
Certificates