missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rabbit Leptin Protein

A cDNA sequence encoding the Leptin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10748-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985048

Quantity Price
1 114.77 € / 20µg
10 107.33 € / 20µg
Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

Leptin Protein
20 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
QSVSSRQRVV GLDFIPGLHP NLSLSTMDQT LAIYQQILAS LPSRNVIQIA NDLENLRDLL HLLASSKSCP LPRASGLETL EGLGGVLEAS
Greater than 95.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor.
Rabbit
Untagged
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3.
Videos
SDS
Documents
Certificates