missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit VCAM1 Protein
A cDNA sequence encoding the VCAM1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1450.00 €
Specifications
Gene ID (Entrez) | 100008901 |
---|---|
Name | VCAM1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | VCAM-1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15995149
|
enQuireBio™
QP13929-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985149
|
enQuireBio™
QP13929-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975149
|
enQuireBio™
QP13929-0.1MG |
100 μg |
1450.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
100008901 | |
Research Use Only | |
VCAM-1 | |
Rabbit | |
His | |
The VCAM1 solution (1 mg/ml) contains 50mM Tris, 300mM NaCl, 10% Glycerol, pH 7.5. |
VCAM1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
AFKIETFPESRSLAQIGDSVSLTCTTTGCASPTFSWRTQIDSPLNGKVRSEGTTSTLTMDPVSFENE HSYLCTATCESKKLEKGIQVEIYSFPKDPEIHLSGPLEVGEPITVKCLVPDVYPFDRLEVDLLKGDYLM KKQDFLEDMDRKSLETKSLEVTFIPVIEDIGKLIVCRAKLHIDEIDSEPKERETTKELQVYISPKNTVIS VNPSTRLQEGGSVTMTCSSEGLPVPEIFWSKKQDNGNLQRLSGNATLTLIAMRMEDSGIYVCEG VNQIGKSRKEVELIVQEKPFTVEISPGPRIAAQIGDPVVLTCSVRGCETPSFSWRTQIDSPLNGQVT SEGTKSLLTLSPVSFENEHSYLCTVTCGHKKLEKGIQVELYSFPRDPEIELSGPPVNGRPVTVSCKVP NVYPFDXLEIELLKGETMMKNKEFLEEEDKKSLETKSLEMTFIPTMEDTGKVLVCQAKLHIDEMEF EPKQRQSTQPLFVNVAPRDIAVWVSPSSIVEEGRSVNMTCSSYGLPAPKILWSRQLKNGDLQPLS ENTTLALISTKLEDSGIYVCEGINLAGKSRKEVELVIQVAPKDIQLTAFPSKSVKEGDTVIISCTCGNV PETWIILKKKAETGDTVLKSIDGAYTIRKAQLEDAGVYECESKNEVGSQLRSITLDVKGRENSKDYF SPE | |
Greater than 90% as determined by SDS-PAGE. |