missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit VEGF / VEGFA / VEGF165 Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) VEGF / VEGFA / VEGF165, was expressed in the hosts and tags indicated.
352.00 € - 3900.00 €
Specifications
Accession Number | NP_999249.1 |
---|---|
Gene ID (Entrez) | 397157 |
Name | VEGF / VEGFA / VEGF165 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036500
|
enQuireBio™
QP6879-YE-100UG |
100 μg |
1050.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16056500
|
enQuireBio™
QP6879-YE-1MG |
1 mg |
3900.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046500
|
enQuireBio™
QP6879-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066500
|
enQuireBio™
QP6879-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076500
|
enQuireBio™
QP6879-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086500
|
enQuireBio™
QP6879-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_999249.1 | |
VEGF / VEGFA / VEGF165 Protein | |
Not determined. | |
Full Length | |
Rabbit | |
His | |
Tris-based buffer, 50% glycerol |
397157 | |
Research Use Only | |
VEGFA | |
Recombinant Protein | |
Yeast | |
YLWLLHAPLL HVSLDSLVAA FSVVFEVLPS SLDIPGSKKE EQASTQLGGV EEEHEASGVV TRQLAFVVDA ITNPTLEKET KTITRIHKAP KFPSHFGFWK RAEEERGGKS SGEKSRKTDG VGERAQAGEQ REGPGQRAAA LTDRQTDTAP SPSAHLLPGR RPTVDAAASG GQQPEPAPEG GVEGVGARGI ALKLFVQLLG CSRSGGVVVR AGGAEPSGAA RSVSSGREEP PPPPPEEEGE EEGEKEEERG PRWRLGAGEP GSWTGEAAVC ADSAPAARAP QALARASAPG ARGARGGAEE SGPSRSPSRR GSASRAGPGR ASETMNFLLS WVHWSLALLL YLHHAKWSQA APMAEEGDNK PHEVVKFMEV YRRSYCQPIE TLVDIFQEYP DEIEYIFKPS CVPLVRCGGC CNDESLECVP TEEFNVTMQI MRIKPHQGQH IGEMSFLQHN KCECRCDKPR R | |
Greater than 90% as determined by SDS-PAGE. |