missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat ACRP30 Protein

A cDNA sequence encoding the ACRP30 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10948-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985658

  • 6549.36 € / 1mg

Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

246253
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Adipoq
Rat
Untagged
Filtered (0.4 microm) and lyophilized in 20 mM Tris buffer, 50 mM NaCl, pH 7.5.
ACRP30 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
HEK 293
GITATEGPGA LVPPPKETCA GWMAGIPGYP GHNGIPGRDG RDGTPGEKGE KGDAGVLGPK GDPGDAGMTG AEGPRGFPGT PGRKGEPGEA AYMYHSAFSV GLETRVTVPN VPIRFTKIFY NQQNHYDGST GKFHCNIPGL YYFSYHITVY MKDVKVSLFK KDKAVLFTYD QYQEKNVDQA SGSMLLHLEV GDQVWLQVYG EGDNNGLYAD NVNDSTFTGF LLYHDTNAAA DYKDDDDK
Videos
SDS
Documents
Certificates