missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat BD 3 Protein
A cDNA sequence encoding the BD 3 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 641623 |
---|---|
Name | BD 3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Defb3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15994158
|
enQuireBio™
QP10533-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984158
|
enQuireBio™
QP10533-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974158
|
enQuireBio™
QP10533-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
641623 | |
Research Use Only | |
Defb3 | |
Recombinant Protein | |
E. coli | |
KKVYNAVSCM TNGGICWLKC SGTFREIGSC GTRQLKCCKK K | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
BD 3 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Measured by its antimicrobial activity against E. coli. The ED50 for this effect is typically 4-20microg/ml, corresponding to a specific activity of 50,000-250,000units/mg. | |
Rat | |
Untagged | |
BD-3 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |