missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat bFGF / FGF2 Protein
Recombinant Protein
107.00 € - 1885.00 €
Specifications
Accession Number | NP_062178.1 |
---|---|
Gene ID (Entrez) | 54250 |
Name | bFGF / FGF2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15996239
|
enQuireBio™
QP5493-10UG |
10 μg |
107.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926249
|
enQuireBio™
QP5493-50UG |
50 μg |
252.00 €
50µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986239
|
enQuireBio™
QP5493-100UG |
100 μg |
433.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15916249
|
enQuireBio™
QP5493-500UG |
500 μg |
1110.00 €
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15906249
|
enQuireBio™
QP5493-1MG |
1 mg |
1885.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_062178.1 | |
bFGF / FGF2 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 1 ng/mL in the BALB/c 3T3 proliferation assay. The bioactivity acceptance criteria is set at 1.0 x 10^6 units/mg. | |
Rat | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |
54250 | |
Research Use Only | |
FGF2 | |
Recombinant Protein | |
E. coli | |
MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS | |
Greater than 95% as determined by SDS-PAGE. |