missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CNTF Protein
A cDNA sequence encoding the CNTF was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11473-1mg
Specifications
13010 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAFAEQT PLTLHRRDLC SRSIWLARKI RSDLTALMES YVKHQGLNKN INLDSVDGVP VASTDRWSEM TEAERLQENL QAYRTFQGML TKLLEDQRVH FTPTEGDFHQ AIHTLMLQVS AFAYQLEELM VLLEQKIPEN EADGMPATVG DGGLFEKKLW GLKVLQELSQ WTVRSIHDLR VISSHQMGIS ALESHYGAKD KQM | |
Greater than 95.0% as determined by SDS-PAGE. |
CNTF Protein | |
Research Use Only | |
Cst3 | |
Rat | |
His | |
CNTF protein solution (0.5 mg/ml) containing phosphate buffered saline (pH 7.4) and 10% glycerol. |