missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Ephrin-A1 / EFNA1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Ephrin-A1 / EFNA1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5964-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988969

Quantity Price
1 639.01 € / 100µg
5 601.25 € / 100µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

NP_446051.2
Ephrin-A1 / EFNA1 Protein
100 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
94268
Greater than 90% as determined by SDS-PAGE.
Research Use Only
EFNA1
Recombinant Protein
E. coli
ADRHIVFWNS SNPKFREEDY TVHVQLNDYL DIICPHYEDD SVADAAMERY TLYMVEHQEY VTCEPQSKDQ VRWKCNQPSA KHGPEKLSEK FQRFTPFTLG KEFKEGHSYY YISKPIYHQE TQCLKLKVTV NGKITHSPHA HANPQEKRLQ ADDPEVQVLH SIGHS
Videos
SDS
Documents
Certificates