missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Fibroblast growth factor 23 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Fibroblast growth factor 23, was expressed in the hosts and tags indicated.
245.00 € - 2645.00 €
Specifications
Accession Number | NP_570110.1 |
---|---|
Gene ID (Entrez) | 170583 |
Name | Fibroblast growth factor 23 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15919579
|
enQuireBio™
QP6045-YE-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959579
|
enQuireBio™
QP6045-YE-500UG |
500 μg |
2100.00 €
500µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929579
|
enQuireBio™
QP6045-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15949579
|
enQuireBio™
QP6045-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15969579
|
enQuireBio™
QP6045-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15939579
|
enQuireBio™
QP6045-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_570110.1 | |
Fibroblast growth factor 23 Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
170583 | |
Research Use Only | |
Fgf23 | |
Recombinant Protein | |
Yeast | |
YSDTSPLLGS NWGSLTHLYT ATARNSYHLQ IHRDGHVDGT PHQTIYSALM ITSEDAGSVV IIGAMTRRFL CMDLRGNIFG SYHFSPENCR FRQWTLENGY DVYLSPKHHY LVSLGRSKRI FQPGTNPPPF SQFLARRNEV PLLHFYTARP RRHTRSAEDP PERDPLNVLK PRPRATPIPV SCSRELPSAE EGGPAASDPL GVLRRGRGDA RRGAGGTDRC RPFPRFV | |
Greater than 80% as determined by SDS-PAGE. |