missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat GDNF Protein
Recombinant Protein
107.00 € - 7105.00 €
Specifications
Accession Number | NP_062012.1 |
---|---|
Gene ID (Entrez) | 25453 |
Name | GDNF Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15986269
|
enQuireBio™
QP5503-2UG |
2 μg |
107.00 €
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966269
|
enQuireBio™
QP5503-10UG |
10 μg |
252.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956269
|
enQuireBio™
QP5503-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996269
|
enQuireBio™
QP5503-500UG |
500 μg |
4325.00 €
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976269
|
enQuireBio™
QP5503-1MG |
1 mg |
7105.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_062012.1 | |
GDNF Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat GDNF induces C6 cell proliferation (typical ED50 is ?0.1 ug/ml). | |
Rat | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
25453 | |
Research Use Only | |
GDNF | |
Recombinant Protein | |
E. coli | |
MSPDKQAAAL PRRERNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCEAAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDSLV YHILRKHSAK RCGCI | |
Greater than 95% as determined by SDS-PAGE. |