missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IFNG / Interferon Gamma Protein
Recombinant Protein
Brand: enQuireBio™ QP5496-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_620235.1 | |
IFNG / Interferon Gamma Protein | |
Research Use Only | |
IFNG | |
Rat | |
Untagged | |
10 mM sodium phosphate, 100 mM sodium chloride, pH 7.5 |
25712 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MQGTLIESLE SLKNYFNSSS MDAMEGKSLL LDIWRNWQKD GNTKILESQI ISFYLRLFEV LKDNQAISNN ISVIESHLIT NFFSNSKAKK DAFMSIAKFE VNNPQIQHKA VNELIRVIHQ LSPESSLRKR KRSRC | |
Greater than 95% as determined by SDS-PAGE. |