missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 1 alpha Protein
A cDNA sequence encoding the IL 1 alpha was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10686-1mg
Specifications
24493 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MAKVPDLFED LKNCYSENEE YSSAIDHLSL NQKSFYDASY GSLHENCTDK FVSLRTSETS KMSTFTFKES RVVVSATSNK GKILKKRRLS FNQPFTEDDL EAIAHDLEET IQPRSAPHSF QNNLRYKLIR IVKQEFIMND SLNQNIYVDM DRIHLKAASL NDLQLEVKFD MYAYSSGGDD SKYPVTLKVS NTQLFVSAQG EDKPVLLKEI PETPKLITGS ETDLIFFWEK INSKNYFTSA AFPELLIATK EQSQVHLARG LPSMIDFQIS | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IL 1 alpha Protein | |
Research Use Only | |
Il1a | |
Rat | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) sterile solution containing 50mM Tris-HCl, pH=8. |