missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 1 alpha Protein
A cDNA sequence encoding the IL 1 alpha was constructed and used to recombinantly synthesize the protein.
131.00 € - 6790.00 €
Specifications
Gene ID (Entrez) | 24493 |
---|---|
Name | IL 1 alpha Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Il1a |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15994648
|
enQuireBio™
QP10686-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974648
|
enQuireBio™
QP10686-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984648
|
enQuireBio™
QP10686-1MG |
1 mg |
6790.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
24493 | |
Research Use Only | |
Il1a | |
Rat | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) sterile solution containing 50mM Tris-HCl, pH=8. |
IL 1 alpha Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MAKVPDLFED LKNCYSENEE YSSAIDHLSL NQKSFYDASY GSLHENCTDK FVSLRTSETS KMSTFTFKES RVVVSATSNK GKILKKRRLS FNQPFTEDDL EAIAHDLEET IQPRSAPHSF QNNLRYKLIR IVKQEFIMND SLNQNIYVDM DRIHLKAASL NDLQLEVKFD MYAYSSGGDD SKYPVTLKVS NTQLFVSAQG EDKPVLLKEI PETPKLITGS ETDLIFFWEK INSKNYFTSA AFPELLIATK EQSQVHLARG LPSMIDFQIS | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |