missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 10 Protein
A cDNA sequence encoding the IL 10 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10688-10ug
Additional Details : Weight : 0.01000kg
Specifications
25325 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by the dose dependent inhibition of MC/9 proliferation is less than 10ng/ml concentration corresponding to a Specific Activity of 100,000IU/mg. | |
Rat | |
Untagged | |
The protein was lyophilized containing 20mM Tris-HCl pH 8.0 and 100mM NaCl. |
IL 10 Protein | |
Research Use Only | |
Il10 | |
Recombinant Protein | |
E. coli | |
SKGHSIRGDN NCTHFPVSQT HMLRELRAAF SQVKTFFQKK DQLDNILLTD SLLQDFKGYL GCQALSEMIK FYLVEVMPQA ENHGPEIKEH LNSLGEKLKT LWIQLRRCHR FLPCENKSKA VEQVKNDFNK LQDKGVYKAM NEFDIFINCI EAYVTLKMKN | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |