missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 15 Protein
A cDNA sequence encoding the IL 15 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10693-10ug
Additional Details : Weight : 0.01000kg
Specifications
25670 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MKILKPYMRN TSILYYLCFL LNSHFLTEAG IHVFILGCVS VGLPKTEANW IDVRYDLEKI ESLIQFIHID TTLYTDSDFH PSCKVTAMNC FLLELQVILH EYSNMTLNET VRNVLYLANS TLSSNKNVIE SGCKECEELE ERNFTEFLQS FIHIVQMFIN TS | |
Greater than 98.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IL 15 Protein | |
Research Use Only | |
Il15 | |
Rat | |
Untagged | |
The protein was lyophilized from 10mM Tris, pH 8.5. |