missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 22 Protein
A cDNA sequence encoding the IL 22 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10703-10ug
Additional Details : Weight : 0.01000kg
Specifications
500836 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The biological activity is measured by its ability to induce IL-10 secretion in COLO 205 (human colon carcinoma cells). The expected ED50 for this effect is 0.15-0.75ng/ml. | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4. |
IL 22 Protein | |
Research Use Only | |
Il22 | |
Recombinant Protein | |
E. coli | |
LPINSQCKLE AANFQQPYIV NRTFMLAKEA SLADNNTDVR LIGEELFRGV KAKDQCYLMK QVLNFTLEDV LLPQSDRFQP YMQEVVPFLT KLSIHLSPCH ISGDDQNIQK NVRQLKETVQ KLGESGEIKA IGELDLLFMS LRNACV | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |