missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 5 Protein
A cDNA sequence encoding the IL 5 was constructed and used to recombinantly synthesize the protein.
131.00 € - 6790.00 €
Specifications
Gene ID (Entrez) | 24497 |
---|---|
Name | IL 5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Il5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954958
|
enQuireBio™
QP10718-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934958
|
enQuireBio™
QP10718-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944958
|
enQuireBio™
QP10718-1MG |
1 mg |
6790.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
24497 | |
Research Use Only | |
Il5 | |
Recombinant Protein | |
E. coli | |
MRMLLCLNVL TLSCVWAIAM EIPMSTVVKE TLIQLSTHRA LLTSNETMRL PVPTHKNHQL CIGEIFQGLD ILKNQTVRGG TVEILFQNLS LIKKYIDGQK EKCGEERRKT RHFLDYLQEF LGVMSTEWAM EV | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IL 5 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 range=0.3-1.0 ng/ml as determined by the dose-dependant stimulation of the proliferation of BCL-1 cells. | |
Rat | |
Untagged | |
Lyophilized from a concentrated (1 mg/ml) solution in water containing no additives. |