missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL17F / IL-17F Protein
Recombinant Protein
Brand: enQuireBio™ QP5515-25ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001015011.2 | |
IL17F / IL-17F Protein | |
Research Use Only | |
IL17F | |
Recombinant Protein | |
E. coli | |
MARRNPKVGL SALQKAGNCP PLEDNSVRVD IRIFNQNQGI SVPRDFQNRS SSPWDYNITR DPDRFPSEIA EAQCRHSGCI NAQGQEDGSM NSVPIQQEIL VLRREPQGCS NSFRLEKMLI KVGCTCVTPI VHHAA | |
Greater than 95% as determined by SDS-PAGE. |
301291 | |
25 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat IL-17F activity was confirmed by measuring IL-6 production from NIH 3T3 cells cultured with varying concentrations of IL-17F. | |
Rat | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |