missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Kcne2 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Kcne2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6254-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_598287.1 | |
Kcne2 Protein | |
Research Use Only | |
Kcne2 | |
Recombinant Protein | |
E. coli | |
MTTLANLTQT LEDAFKKVFI TYMDSWRRNT TAEQQALQAR VDAENFYYVI LYLMVMIGMF AFIVVAILVS TVKSKRREHS QDPYHQYIVE DWQQKYRSQI LHLEDSKATI HENLGATGFT VSP | |
Greater than 90% as determined by SDS-PAGE. |
171138 | |
50 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |