missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat LCN2 Protein
A cDNA sequence encoding the LCN2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12546-2ug
Additional Details : Weight : 0.01000kg
Specifications
170496 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQDSTQNL IPAPPLISVP LQPGFWTERF QGRWFVVGLA ANAVQKERQS RFTMYSTIYE LQEDNSYNVT SILVRGQGCR YWIRTFVPSS RPGQFTLGNI HSYPQIQSYD VQVADTDYDQ FAMVFFQKTS ENKQYFKVTL YGRTKGLSDE LKERFVSFAK SLGLKDNNIV FSVPTDQCID N | |
Greater than 95.0% as determined by SDS-PAGE. |
LCN2 Protein | |
Research Use Only | |
Lcn2 | |
Rat | |
His | |
LCN2 protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 20% glycerol. |