missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat MCP-3 / CCL7 Protein
Recombinant Protein
Brand: enQuireBio™ QP5500-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001007613.1 | |
MCP-3 / CCL7 Protein | |
Research Use Only | |
CCL7 | |
Rat | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
287561 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
QPDGTNSSTC CYVKKQKIPK RNLKSYRKIT SSRCPWEAVI FKTKKGMEVC AEAHQKWVEE AIAYLDMKTS TPKP | |
Greater than 95% as determined by SDS-PAGE. |