missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat MDC Protein
A cDNA sequence encoding the MDC was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | MDC Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | MDC |
Biological Activity | Determined by its ability to chemoattract human activated lymphocytes using a concentration range of 10.0-100.0 ng/ml. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15955158
|
enQuireBio™
QP10783-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945158
|
enQuireBio™
QP10783-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935158
|
enQuireBio™
QP10783-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MDC Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Determined by its ability to chemoattract human activated lymphocytes using a concentration range of 10.0-100.0 ng/ml. | |
Rat | |
Untagged | |
The protein was lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4. |
Research Use Only | |
MDC | |
Recombinant Protein | |
E. coli | |
GPYGANVEDS ICCQDYIRHP LPPRFVKEFY WTSKSCRKPG VVLITIKNRD ICADPRMLWV KKILHKLA | |
Greater than 97.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |