missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Mucosal addressin cell adhesion molecule 1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Mucosal addressin cell adhesion molecule 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6331-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_062190.1 | |
Mucosal addressin cell adhesion molecule 1 Protein | |
Research Use Only | |
Madcam1 | |
Recombinant Protein | |
E. coli | |
QSFQVNPPEP EVAVAMGTSL QINCSMSCDK DIARVHWHGL DTNLGNVQTL PGSRVLSVRG MLSDTGTRVC VGSCGSRSFQ HSVKILVYAF PDQLEVTPEF LVPGRDQVVS CTAHNIWPAG PDSLSFALLR GEQSLEGAQA LETEQEEEMQ ETEGTPLFQV TQRWLLPSLG TPALPALYCQ VTMQLPKLVL THRRKIPVLQ SQTSPEPPST TSAKPYILTS SHTTKAVSTG LSSVALPSTP LSSEGPCYPE IHQNPEADWE LLCEASCGSG VTVHWTLAPG DLAAYHKREA GAQAWLSVLP LGPIPEGWFQ CRMDPGGQVT SLYVTGQVIP NPSS | |
Greater than 90% as determined by SDS-PAGE. |
54266 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |