missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Myosin-binding protein C, cardiac-type Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Myosin-binding protein C, cardiac-type, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9297-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001099960.1 | |
Myosin-binding protein C, cardiac-type Protein | |
Research Use Only | |
Mybpc3 | |
Recombinant Protein | |
Yeast | |
PKIHLDCPGS TPDTIVVVAG NKLRLDVPIS GDPAPTVIWQ KTITQGKKAS AGPPPGAPED AGADEEWVFD KKLLCETEGR VRVETTKDRS VFTVEGAEKE DEGVYTVTVK NPVGEDQVNL TVKVIDVPDA PAAPKISNVG EDSCIVQWEP PAYDGGQPVL GYILERKKKK SYRWMRLNFD LLRELSHEAR RMIEGVAYEM RVYAVNAVGM SRPSPASQPF | |
Greater than 90% as determined by SDS-PAGE. |
295929 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |