missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Nitric oxide synthase, endothelial Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Nitric oxide synthase, endothelial, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_068610.1 |
---|---|
Gene ID (Entrez) | 24600 |
Name | Nitric oxide synthase, endothelial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16016891
|
enQuireBio™
QP8865-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036891
|
enQuireBio™
QP8865-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026891
|
enQuireBio™
QP8865-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046891
|
enQuireBio™
QP8865-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056891
|
enQuireBio™
QP8865-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066891
|
enQuireBio™
QP8865-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_068610.1 | |
Nitric oxide synthase, endothelial Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
24600 | |
Research Use Only | |
Nos3 | |
Recombinant Protein | |
E. coli | |
ATILYGSETG RAQSYAQQLG RLFRKAFDPR VLCMDEYDVV SLEHEALVLV VTSTFGNGDP PENGESFAAA LMEMSGPYNS SPRPEQHKSY KIRFNSVSCS DPLVSSWRRK RKESSNTDSA GALGTLRFCV FGLGSRAYPH FCAFARAVDT RLEELGGERL LQLGQGDELC GQ | |
Greater than 90% as determined by SDS-PAGE. |