missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Nqo2 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Nqo2, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_001004214.1 |
---|---|
Gene ID (Entrez) | 291084 |
Name | Nqo2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16040291
|
enQuireBio™
QP7703-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060291
|
enQuireBio™
QP7703-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16050291
|
enQuireBio™
QP7703-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070291
|
enQuireBio™
QP7703-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080291
|
enQuireBio™
QP7703-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16090291
|
enQuireBio™
QP7703-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001004214.1 | |
Nqo2 Protein | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
291084 | |
Research Use Only | |
Nqo2 | |
Recombinant Protein | |
E. coli | |
MAGKKVLLVY AHQEPKSFNG SMKQVAVEEL SKQGCTVTVS DLYTMNFEPR ATRNDVTGAL SNPEVFKYGI EAYEAYKKKA LTSDILEEQR KVQEADLVIF QFPLYWFSVP AILKGWMDRV LCQGFAFDVP GFYDSGFLKD KLALLSFTTG GTAEMYTKAG VNGDFRYFLW PLQHGTLHFC GFKVLAPQIS FGPEVSSEEQ RKVMLASWVQ RLKSIWKEEP IHCTPSWYFQ G | |
Greater than 90% as determined by SDS-PAGE. |