missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Nqo2 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Nqo2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7703-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001004214.1 | |
Nqo2 Protein | |
500 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
291084 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Nqo2 | |
Recombinant Protein | |
Yeast | |
MAGKKVLLVY AHQEPKSFNG SMKQVAVEEL SKQGCTVTVS DLYTMNFEPR ATRNDVTGAL SNPEVFKYGI EAYEAYKKKA LTSDILEEQR KVQEADLVIF QFPLYWFSVP AILKGWMDRV LCQGFAFDVP GFYDSGFLKD KLALLSFTTG GTAEMYTKAG VNGDFRYFLW PLQHGTLHFC GFKVLAPQIS FGPEVSSEEQ RKVMLASWVQ RLKSIWKEEP IHCTPSWYFQ G |