missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Osteocalcin Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Osteocalcin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5717-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_038200.1 | |
Osteocalcin Protein | |
Research Use Only | |
Bglap | |
Recombinant Protein | |
E. coli | |
YLNNGLGAPA PYPDPLEPHR EVCELNPNCD ELADHIGFQD AYKRIYGTTV | |
Greater than 82.2% as determined by SDS-PAGE. |
25295 | |
200 μg | |
Not determined. | |
Full Length | |
Rat | |
GST | |
Tris-based buffer, 50% glycerol |