missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PFN1 Protein
A cDNA sequence encoding the PFN1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | PFN1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Rat |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15942269
|
enQuireBio™
QP13020-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932269
|
enQuireBio™
QP13020-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922269
|
enQuireBio™
QP13020-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PFN1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
His | |
PFN1 protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAGWNA YIDSLMADGT CQDAAIVGYK DSPSVWAAVP GKTFVSITPA EVGVLVGKDR SSFFVNGLTL GGQKCSVIRD SLLQDGEFTM DLRTKSTGGA PTFNVTVTMT AKTLVLLMGK EGVHGGLINK KCYEMASHLR RSQY | |
Greater than 95.0% as determined by SDS-PAGE. |