missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat SDF 1b Protein
A cDNA sequence encoding the SDF 1b was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10863-2ug
Additional Details : Weight : 0.01000kg
Specifications
24772 | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
Cxcl12 | |
Recombinant Protein | |
E. coli | |
KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKSNNRQVC IDPKLKWIQE YLDKALNKRL KM |
SDF 1b Protein | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The specific activity as determined by its ability to chemoattract human peripheral blood monocytes at 100-150ng/ml corresponding to a Specific Activity of 6,667-10,000IU/mg. | |
Rat | |
Untagged | |
The Rat SDF1B protein was lyophilized from a concentrated (1 mg/ml) sterile solution containing 20mM Phsophate buffer pH 7.4 and 0.15M NaCl. |