missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat SerpinA1 / A1AT Protein
A DNA sequence encoding the Rattus norvegicus (Rat) SerpinA1 / A1AT, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6666-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_071964.2 | |
SerpinA1 / A1AT Protein | |
Research Use Only | |
Serpina1 | |
Recombinant Protein | |
Yeast | |
EDAQETDTSQ QDQSPTYRKI SSNLADFAFS LYRELVHQSN TSNIFFSPMS ITTAFAMLSL GSKGDTRKQI LEGLEFNLTQ IPEADIHKAF HHLLQTLNRP DSELQLNTGN GLFVNKNLKL VEKFLEEVKN NYHSEAFSVN FADSEEAKKV INDYVEKGTQ GKIVDLMKQL DEDTVFALVN YIFFKGKWKR PFNPEHTRDA DFHVDKSTTV KVPMMNRLGM FDMHYCSTLS SWVLMMDYLG NATAIFLLPD DGKMQHLEQT LTKDLISRFL LNRQTRSAIL YFPKLSISGT YNLKTLLSSL GITRVFNNDA DLSGITEDAP LKLSQAVHKA VLTLDERGTE AAGATVVEAV PMSLPPQVKF DHPFIFMIVE SETQSPLFVG KVIDPTR | |
Greater than 90% as determined by SDS-PAGE. |
24648 | |
10 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |