missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus E.coli Enoyl-ACP Reductase / FABI Protein
A DNA sequence encoding the Staphylococcus aureus (strain MRSA252) E.coli Enoyl-ACP Reductase / FABI, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | WP_000933195.1 |
---|---|
Gene ID (Entrez) | 31213761 |
Name | S. aureus E.coli Enoyl-ACP Reductase / FABI Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16030591
|
enQuireBio™
QP7753-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16050591
|
enQuireBio™
QP7753-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16040591
|
enQuireBio™
QP7753-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060591
|
enQuireBio™
QP7753-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070591
|
enQuireBio™
QP7753-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080591
|
enQuireBio™
QP7753-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_000933195.1 | |
S. aureus E.coli Enoyl-ACP Reductase / FABI Protein | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
31213761 | |
Research Use Only | |
fabI | |
Recombinant Protein | |
E. coli | |
MLNLENKTYV IMGIANKRSI AFGVAKVLDQ LGAKLVFTYR KERSRKELEK LLEQLNQPEA HLYQIDVQSD EEVINGFEQI GKDVGNIDGV YHSIAFANME DLRGRFSETS REGFLLAQDI SSYSLTIVAH EAKKLMPEGG SIVATTYLGG EFAVQNYNVM GVAKASLEAN VKYLALDLGP DNIRVNAISA GPIRTLSAKG VGGFNTILKE IEERAPLKRN VDQVEVGKTA AYLLSDLSSG VTGENIHVDS GFHAIK | |
Greater than 90% as determined by SDS-PAGE. |