missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus esxA Protein
A DNA sequence encoding the Staphylococcus aureus (strain MSSA476) esxA, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | WP_001240826.1 |
---|---|
Gene ID (Entrez) | 31213083 |
Name | S. aureus esxA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16020721
|
enQuireBio™
QP7776-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16040721
|
enQuireBio™
QP7776-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16030721
|
enQuireBio™
QP7776-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050721
|
enQuireBio™
QP7776-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060721
|
enQuireBio™
QP7776-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070721
|
enQuireBio™
QP7776-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_001240826.1 | |
S. aureus esxA Protein | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
31213083 | |
Research Use Only | |
esxA | |
Recombinant Protein | |
E. coli | |
MAMIKMSPEE IRAKSQSYGQ GSDQIRQILS DLTRAQGEIA ANWEGQAFSR FEEQFQQLSP KVEKFAQLLE EIKQQLNSTA DAVQEQDQQL SNNFGLQ | |
Greater than 90% as determined by SDS-PAGE. |