missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Staphopain B Protein

A DNA sequence encoding the Staphylococcus aureus Staphopain B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9880-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16058901

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_001088793.1
50 μg
Not determined.
Full Length
S. aureus
His
Tris-based buffer, 50% glycerol
S. aureus Staphopain B Protein
Research Use Only
sspB
Recombinant Protein
Yeast
DQVQYENTLK NFKIREQQFD NSWCAGFSMA ALLNATKNTD TYNAHDIMRT LYPEVSEQDL PNCSTFPNQM IEYGKSQGRD IHYQEGVPSY EQVDQLTKDN VGIMILAQSV SQNPNDPHLG HALAVVGNAK INDQEKLIYW NPWDTELSIQ DADSSLLHLS FNRDYNWYGS MIGY
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions