missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Soybean Soybean P34 GST Protein
A cDNA sequence encoding the Soybean P34 GST was constructed and used to recombinantly synthesize the protein.
228.00 € - 1740.00 €
Specifications
Name | P34 GST Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Soybean |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15923989
|
enQuireBio™
QP13564-100UG |
100 μg |
228.00 €
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943989
|
enQuireBio™
QP13564-500UG |
500 μg |
870.00 €
500µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933989
|
enQuireBio™
QP13564-1MG |
1 mg |
1740.00 €
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
P34 GST Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Soybean | |
GST | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, pH 8.0, 60mM NaCl, 10mM glutathione and 50% glycerol. |