missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Toxoplasma Gondii T. gondii p24 Protein
A cDNA sequence encoding the T. gondii p24 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13664-1mg
Additional Details : Weight : 0.01000kg
Specifications
Toxoplasma Gondii T. gondii p24 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
(1 mg/ml) 50mM Tris pH 8.0, 1.5M urea and 50% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Toxoplasma Gondii | |
Untagged | |
Toxoplasma protein is >95% pure as determined by 10% PAGE (coomassie staining). |