missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Toxoplasma Gondii T. gondii p24 Protein
A cDNA sequence encoding the T. gondii p24 was constructed and used to recombinantly synthesize the protein.
228.00 € - 1450.00 €
Specifications
Name | Toxoplasma Gondii T. gondii p24 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Toxoplasma Gondii |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974309
|
enQuireBio™
QP13664-100UG |
100 μg |
228.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15994309
|
enQuireBio™
QP13664-500UG |
500 μg |
870.00 €
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984309
|
enQuireBio™
QP13664-1MG |
1 mg |
1450.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Toxoplasma Gondii T. gondii p24 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Toxoplasma Gondii | |
Untagged | |
Toxoplasma protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
(1 mg/ml) 50mM Tris pH 8.0, 1.5M urea and 50% glycerol. |