missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Toxoplasma Gondii T. gondii p40 Protein
A cDNA sequence encoding the T. gondii p40 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13667-500ug
Additional Details : Weight : 0.01000kg
Specifications
Toxoplasma Gondii T. gondii p40 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
1x PBS and 25mM arginine. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Toxoplasma Gondii | |
His | |
Toxoplasma protein is >95% pure as determined by 10% PAGE (coomassie staining). |