missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Varicella Zoster Virus VZV ORF9 Protein
A cDNA sequence encoding the VZV ORF9 was constructed and used to recombinantly synthesize the protein.
228.00 € - 1450.00 €
Specifications
Name | Varicella Zoster Virus VZV ORF9 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Varicella Zoster Virus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15965219
|
enQuireBio™
QP13952-100UG |
100 μg |
228.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985219
|
enQuireBio™
QP13952-500UG |
500 μg |
870.00 €
500µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975219
|
enQuireBio™
QP13952-1MG |
1 mg |
1450.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Varicella Zoster Virus VZV ORF9 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Varicella Zoster Virus | |
Untagged | |
Varicella protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-Hcl pH 8.0, 1mM EDTA and 50% glycerol. |